missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97992-100ul
This item is not returnable.
View return policy
Description
SLC22A7 Polyclonal antibody specifically detects SLC22A7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC22A7 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| hOAT2, liver-specific transporter, NLTMGC45202, Novel liver transporter, OAT2MGC24091, Organic anion transporter 2, solute carrier family 22 (organic anion transporter), member 7, solute carrier family 22 member 7 | |
| Produced in rabbits immunized with E. coli-derived Human SLC22A7 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10864 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction