missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RWDD4 Protein
A cDNA sequence encoding the RWDD4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13377-10ug
Additional Details : Weight : 0.01000kg
Specifications
RWDD4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSANEDQ EMELEALRSI YEGDESFREL SPVSFQYRIG ENGDPKAFLI EISWTETYPQ TPPILSMNAF FNNTISSAVK QSILAKLQEA VEANLGTAMT YTLFEYAKDN KEQFMENHNP INSATSISNI ISIETPNTAP SSKKKDKKEQ LSKAQKRKLA DKTDHKGELP RGWNWVDVVK HLSKTGSKDD E | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RWDD4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |