missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH2D1B Protein
A cDNA sequence encoding the SH2D1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13486-10ug
Additional Details : Weight : 0.01000kg
Specifications
SH2D1B Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMDLPYYH GRLTKQDCET LLLKEGVDGN FLLRDSESIP GVLCLCVSFK NIVYTYRIFR EKHGYYRIQT AEGSPKQVFP SLKELISKFE KPNQGMVVHL LKPIKRTSPS LRWRGLKLEL ETFVNSNSDY VDVLPHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SH2D1B protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |