missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SH2D1B Protein

A cDNA sequence encoding the SH2D1B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13486-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923729

  • 7200.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

SH2D1B Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SH2D1B protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
ADPMDLPYYH GRLTKQDCET LLLKEGVDGN FLLRDSESIP GVLCLCVSFK NIVYTYRIFR EKHGYYRIQT AEGSPKQVFP SLKELISKFE KPNQGMVVHL LKPIKRTSPS LRWRGLKLEL ETFVNSNSDY VDVLPHHHHH H
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions