missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TIGIT (A103) Fc Chimera Protein

Product Code. 18707146
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18707146 1 mg 1mg
18116800 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18707146 Supplier R&D Systems™ Supplier No. 7898TGB01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR (Catalog # 2530-CD ) is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Human TIGIT Fc Chimera that produces 50% optimal binding response is 40-200 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAI01289
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 201633
Molecular Weight (g/mol) M.W. (Observed): 40-53 kDa, reducing conditions; M.W. (theoretical): 40 kDa
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human TIGIT protein Human TIGIT (Met22-Pro141) (N-terminus) Accession # AAI01289 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity TIGIT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TIGIT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.