missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tripartite motif-containing protein 72 Protein

A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 72, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10131-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15912888

  • 1690.00€ / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001008275.2
Tripartite motif-containing protein 72 Protein
Research Use Only
TRIM72
Recombinant Protein
Mammal
MSAAPGLLHQ ELSCPLCLQL FDAPVTAECG HSFCRACLGR VAGEPAADGT VLCPCCQAPT RPQALSTNLQ LARLVEGLAQ VPQGHCEEHL DPLSIYCEQD RALVCGVCAS LGSHRGHRLL PAAEAHARLK TQLPQQKLQL QEACMRKEKS VAVLEHQLVE VEETVRQFRG AVGEQLGKMR VFLAALEGSL DREAERVRGE AGVALRRELG SLNSYLEQLR QMEKVLEEVA DKPQTEFLMK YCLVTSRLQK ILAESPPPAR LDIQLPIIS
Greater than 90% as determined by SDS-PAGE.
493829
100 μg
Not determined.
Partial (see sequence information for more details).
Human
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions