missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tripartite motif-containing protein 72 Protein

A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 72, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10131-ma-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932888

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_001008275.2
Tripartite motif-containing protein 72 Protein
Research Use Only
TRIM72
Recombinant Protein
Mammal
MSAAPGLLHQ ELSCPLCLQL FDAPVTAECG HSFCRACLGR VAGEPAADGT VLCPCCQAPT RPQALSTNLQ LARLVEGLAQ VPQGHCEEHL DPLSIYCEQD RALVCGVCAS LGSHRGHRLL PAAEAHARLK TQLPQQKLQL QEACMRKEKS VAVLEHQLVE VEETVRQFRG AVGEQLGKMR VFLAALEGSL DREAERVRGE AGVALRRELG SLNSYLEQLR QMEKVLEEVA DKPQTEFLMK YCLVTSRLQK ILAESPPPAR LDIQLPIIS
Greater than 90% as determined by SDS-PAGE.
493829
50 μg
Not determined.
Partial (see sequence information for more details).
Human
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions