Learn More
Cytiva Recombinant Taq DNA Polymerase
Product Code. 11349951 Shop All Cytiva Products
Change view
Click to view available options
:
5000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11349951 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11349951 Supplier Cytiva Supplier No. 28937346
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Taq DNA Polymerase (cloned) is the recombinant form of the native enzyme from Thermus aquaticus expressed in E. coli.
  • Highly purified recombinant enzyme
  • Excellent batch-to-batch reproducibility
  • Each lot is tested to be free of nuclease contamination