missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza-B Tokio Protein

A cDNA sequence encoding the Influenza-B Tokio was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12443-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920439

  • 3030.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza-B Tokio Protein
Research Use Only
Recombinant Protein
Influenza B virus
The B/Tokio/53/99solution (1.7 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers