missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cyclophilin B Protein

A cDNA sequence encoding the Cyclophilin B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10569-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934278

  • 125.00€ / 2µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

19035
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Ppib
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNDKKKG PKVTVKVYFD LQIGDESVGR VVFGLFGKTV PKTVDNFVAL ATGEKGFGYK NSKFHRVIKD FMIQGGDFTR GDGTGGKSIY GERFPDENFK LKHYGPGWVS MANAGKDTNG SQFFITTVKT SWLDGKHVVF GKVLEGMDVV RKVESTKTDSRDKPLKDVII VDSGKIEVEK PFAIAKE
Cyclophilin B Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 1,000 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmole of suc-AAFP-PNA per minute at 37C in Tris€“HCl pH 8.0 using chymotrypsin.
Mouse
His
Cyclophilin B protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions