missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse GM-CSF Protein

Product Code. 18796686
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796686 5 μg 5µg
18777476 1 mg 1mg
18746446 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18796686 Supplier R&D Systems™ Supplier No. 415ML005/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse GM-CSF Protein is derived from E. coli. The Recombinant Mouse GM-CSF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14AD9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1437
Molecular Weight (g/mol) 14.3 kDa
Quantity 5 μg
Source <EM>E. coli</EM>-derived mouse GM-CSF protein Ala18-Lys141, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Gene Symbol Csf2
Conjugate Unconjugated
Cross Reactivity GM-CSF
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2μm filtered solution in PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.