missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A6 Protein
A cDNA sequence encoding the S100A6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13397-10ug
Additional Details : Weight : 0.01000kg
Specifications
20200 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MACPLDQAIG LLVAIFHKYS GKEGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMDDL DRNKDQEVNF QEYVAFLGAL ALIYNEALK | |
Greater than 90% as determined by SDS-PAGE. |
S100A6 Protein | |
Research Use Only | |
S100a6 | |
Mouse | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing no additives. |