missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse WIF-1 Protein

Product Code. 18357747
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357747 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357747 Supplier R&D Systems™ Supplier No. 135WF050
  • ONLINE EXCLUSIVE PRICE
    467.00€
    441.00€ / 50µg
    Save 26.00€
    5% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse WIF-1 Protein is derived from NS0. The Recombinant Mouse WIF-1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetic Acid, NaCl, PEG3350 and CHAPS, pH5.0.
Gene ID (Entrez) 24117
Molecular Weight (g/mol) 41.1 kDa
Name WIF-1
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse WIF-1 protein, (N-terminus) Mouse WIF-1 (Gly29-Trp379) Accession &Num; Q9WUA1 HIEGR GGGSGGGSGGGS 10-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.