missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Batroxobin Protein
A cDNA sequence encoding the Batroxobin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11130-1mg
Additional Details : Weight : 0.01000kg
Specifications
Batroxobin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVLIRVIANL LILQVSYAQK SSELVIGGDE CDINEHPFLA FMYYSPRYFC GMTLINQEWV LTAAHCNRRF MRIHLGKHAG SVANYDEVVR YPKEKFICPN KKKNVITDKD IMLIRLDRPV KNSEHIAPLS LPSNPPSVGS VCRIMGWGAI TTSEDTYPDV PHCANINLFN NTVCREAYNG LPAKTLCAGV LQGGIDTCGG DSGGPLICNG QFQGILSWGS DPCAEPRKPA FYTKVFDYLP WIQSIIAGNK TATCP |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |