missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PROK Tritirachium album Protein
A cDNA sequence encoding the PROK Tritirachium album was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10849-20mg
Additional Details : Weight : 0.01000kg
Specifications
PROK Tritirachium album Protein | |
Research Use Only | |
PROK | |
Recombinant Protein | |
Yeast | |
MRLSVLLSLL PLALGAPAVE QRSEAAPLIE ARGEMVANKY IVKFKEGSAL SALDAAMEKI SGKPDHVYKN VFSGFAATLD ENMVRVLRAH PDVEYIEQDA VVTINAAQTN APWGLARISS TSPGTSTYYY DESAGQGSCV YVIDTGIEAS HPEFEGRAQM VKTYYYSSRD GNGHGTHCAG TVGSRTYGVA KKTQLFGVKV LDDNGSGQYS TIIAGMDFVA SDKNNRNCPK GVVASLSLGG GYSSSVNSAA ARLQSSGVMV AVAAGNNNAD ARNYSPASEP SVCTVGASDR YDRRSSFSNY GSVLDIFGPG TSILSTWIGG STRSISGTSM ATPHVAGLAA YLMTLGKTTA ASACRYIADT ANKGDLSNIP FGTVNLLAYN NYQA | |
Greater than 99% as determined by SDS-PAGE. |
20 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
34 Units/mg protein. One unit is defined as the amount of enzyme that will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 mol tyrosine per min at 37C, pH 7.5 (color by Folin-Ciocalteu reagent). | |
Other | |
Untagged | |
The Proteinase-K was lyophilized without any additives. |