missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other RNASEH1 Protein

A cDNA sequence encoding the RNASEH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13325-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923219

  • 189.00€ / 20µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

RNASEH1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
RNASEH1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 2mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLKQVEI FTDGSCLGNP GPGGYGAILR YRGREKTFSA GYTRTTNNRM ELMAAIVALE ALKEHCEVIL STDSQYVRQG ITQWIHNWKK RGWKTADKKP VKNVDLWQRL DAALGQHQIK WEWVKGHAGH PENERCDELA RAAAMNPTLE DTGYQVEV
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions