missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other UPP1ella Protein

A cDNA sequence encoding the UPP1ella was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13901-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945069

  • 2120.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UPP1ella Protein
Research Use Only
30 U/mg protein.
Other
Untagged
The UPase was lyophilized from 1 mg/ml solution containing 25mM Tris-HCl, pH 8.0, 0.15M NaCl.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSKSDVFHLG LTKNDLQGAQ LAIVPGDPER VEKIAALMDK PVKLASHREF TSWRAELDGK AVIVCSTGIG GPSTSIAVEE LAQLGIRTFL RIGTTGAIQP HINVGDVLVT TASVRLDGAS LHFAPMEFPA VADFACTTAL VEAAKSIGAT THVGVTASSD TFYPGQERYD TYSGRVVRRF KGSMEEWQAM GVMNYEMESA TLLTMCASQG LRAGMVAGVI VNRTQQEIPN AETMKQTESH AVKIVVEAAR RLL
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions