missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Prolactin Antagonist Protein

A cDNA sequence encoding the Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP14010-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925409

  • 1880.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Prolactin Antagonist Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3.
Greater than 99.0% as determined by SDS-PAGE.
Research Use Only
Ovine Prolactin Antagonist is devoid of agonistic activity and capable of inhibiting biological activity of Ovine Prolactin or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells.
Ovine
His
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions