missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat FABP3 / H-FABP Protein

A DNA sequence encoding the Rattus norvegicus (Rat) FABP3 / H-FABP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6013-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989359

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_077076.1
FABP3 / H-FABP Protein
Research Use Only
Fabp3
Recombinant Protein
E. coli
ADAFVGTWKL VDSKNFDDYM KSLGVGFATR QVASMTKPTT IIEKNGDTIT IKTHSTFKNT EISFQLGVEF DEVTADDRKV KSVVTLDGGK LVHVQKWDGQ ETTLTRELSD GKLILTLTHG NVVSTRTYEK EA
Greater than 90% as determined by SDS-PAGE.
79131
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions