missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FABP3 / H-FABP Protein
A DNA sequence encoding the Rattus norvegicus (Rat) FABP3 / H-FABP, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_077076.1 |
---|---|
Gene ID (Entrez) | 79131 |
Name | FABP3 / H-FABP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959359
|
enQuireBio™
QP6013-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979359
|
enQuireBio™
QP6013-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969359
|
enQuireBio™
QP6013-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15989359
|
enQuireBio™
QP6013-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15999359
|
enQuireBio™
QP6013-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15909369
|
enQuireBio™
QP6013-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_077076.1 | |
FABP3 / H-FABP Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79131 | |
Research Use Only | |
Fabp3 | |
Recombinant Protein | |
E. coli | |
ADAFVGTWKL VDSKNFDDYM KSLGVGFATR QVASMTKPTT IIEKNGDTIT IKTHSTFKNT EISFQLGVEF DEVTADDRKV KSVVTLDGGK LVHVQKWDGQ ETTLTRELSD GKLILTLTHG NVVSTRTYEK EA | |
Greater than 90% as determined by SDS-PAGE. |