missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat FGF 21 Protein

A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10600-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924378

  • 189.00€ / 20µg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

FGF 21 Protein
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
AYPISDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGTAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GTLYGSPHFD PEACSFRELL LKDGYNVYQS EAHGLPLRLP QKDSQDPATR GPVRFLPMPG LPHEPQEQPG VLPPEPPDVG SSDPLSMVEP LQGRSPSYAS
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The ED50 determined by a cell proliferation assay using murine NIH/3T3 cells is less than 700 ng/ml, corresponding to a specific activity of > 1.4 M 1000 IU/mg in the presence of 5microg/ml of rMuKlotho-beta.
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions