missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus HTNV Protein

A cDNA sequence encoding the HTNV was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12359-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930169

  • 832.00€ / 500µg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Virus HTNV Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Virus
GST
HTNV protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
HTNV (5 mg/ml) is formulated in 1xPBS pH 7.4 and 0.5mM EDTA.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions