missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus HTNV Protein
A cDNA sequence encoding the HTNV was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12359-1mg
Additional Details : Weight : 0.01000kg
Specifications
Virus HTNV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HTNV (5 mg/ml) is formulated in 1xPBS pH 7.4 and 0.5mM EDTA. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
GST | |
HTNV protein is >95% pure as determined by 10% PAGE (coomassie staining). |