Learn More
Agencinox™ Rectangular Tray
Product Code. 11741933
Change view
Click to view available options
Dimensions (L x W x H):
170 x 110 x 20 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11741933 170 x 110 x 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11741933 Supplier Agencinox™ Supplier No. 1217
In Stock
Add to basket
Edge
Add to basket
Use these trays for a variety of applications. Agencinox™ Rectangular Trays ideal for use in medical environments.
  • Rectangular tray
  • Stainless steel construction
  • Available in a variety of sizes
Specifications
Dimensions (L x W) 170 x 110 mm
Dimensions (L x W x H) 170 x 110 x 20 mm
Height (Metric) 20 mm
Length (Metric) 170 mm
Material Stainless Steel
Type Tray
Width (Metric) 110 mm
For Use With Instruments
Shape Rectangular