missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Red-Gal
Description
An alternative chromogenic substrate for β-D-galactosidase.
Red-GAL is used in conjuction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colorimetric assay to detect recombinants from non-recombinants.
- Substance Name: 6-Chloro-3-indolyl-β-D-galactopyranoside
- CAS Number: 138182-21-5
- Molecular Formula: C14H16CINO6
- Molecular Weight: 329.7
Specifications
Specifications
| Quantity | 1 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?