Learn More
Abeba™ Reflexor™ 36610 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 36610/39
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 15630398
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • Integrated Reflexor™ massage footbed
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012
Microfiber
39
Polyurethane
Black
Leather
Slip-resistant
36610