Learn More
Abeba™ Reflexor™ 36610 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 36610/39
Additional Details : Weight : 0.70000kg
Description
- CE EN ISO 20347:2012
- Smooth leather
- Black
- Slip resistance PU outsole sole
- Integrated Reflexor™ massage footbed
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 | |
Microfiber | |
39 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
36610 |