Learn More
Abeba™ Reflexor™ 36900 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 36900/36
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15630788
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRB
Microfiber
36
Polyurethane
White
Leather
Slip-resistant
36900