missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg3G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06426-100ul
This item is not returnable.
View return policy
Description
Reg3G Polyclonal antibody specifically detects Reg3G in Human samples. It is validated for ELISA
Specifications
| Reg3G | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| LPPM429, MGC118999, MGC119001, Pancreatitis-associated protein 1B, Pancreatitis-associated protein IB, PAP IB, PAP-1B, PAP1BMGC118998, PAPIB, Reg III-gamma, REG-3-gamma, regenerating gene III, regenerating islet-derived 3 gamma, regenerating islet-derived protein 3-gamma, Regenerating islet-derived protein III-gamma, REG-III, UNQ429 | |
| Produced in rabbits immunized with purified, recombinant Human Reg3G (Accession#: NP_001008388.1; Met 1-Asp 175) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 130120 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction