Learn More
Reichardt’s Dye (Technical Grade), TRC
Product Code. 30136471
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136471 250 mg 250mg
30006635 100 mg 100mg
30010686 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136471 Supplier Toronto Research Chemicals Supplier No. TRCR225240250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10081-39-7
Molecular Formula C41H29NO
Molecular Weight (g/mol) 551.68
Synonym Pyridinium, 1-(2'-hydroxy[1,1':3',1' '-terphenyl]-5'-yl)-2,4,6-triphenyl-, inner salt (9CI, ACI), 1-(2'-Hydroxy-m-terphenyl-5'-yl)-2,4,6-triphenylpyridinium hydroxide, inner salt (7CI), Pyridinium, 1-(3,5-diphenyl-4-hydroxyphenyl)-2,4,6-triphenyl-, hydroxide, inner salt (8CI), 1-(2'-Hydroxy[1,1':3',1' '-terphenyl]-5'-yl)-2,4,6-triphenylpyridinium hydroxide inner salt, 2,6-Diphenyl-4-(2,4,6-triphenyl-1-pyridinio)phenolate, 2,6-Diphenyl-4-(2,4,6-triphenyl-1-pyridinio)phenoxide, 2,6-Diphenyl-4-(2,4,6-triphenylpyridin-1-ium-1-yl)phenolate, 2,6-Diphenyl-4-(2,4,6-triphenylpyridinium-1-yl)phenolate, 2,6-Diphenyl-4-(2',4',6'-triphenyl-1-pyridinium)phenolate, 4-(2,4,6-Triphenylpyridinio)-2,6-diphenylphenoxide, 4-(2,4,6-Triphenylpyridinium-1-yl)-2,6-diphenylphenolate, Betaine 30, Dimroth-Reichardt betaine dye, ET 30, Reichard's ET 30 dye, Reichardt dye, Reichardt's betain, Reichardt's dye, Reichardt's ET(30) betaine dye
IUPAC Name 2,6-diphenyl-4-(2,4,6-triphenylpyridin-1-ium-1-yl)phenolate
SMILES [O-]c1c(cc(cc1c2ccccc2)[n+]3c(cc(cc3c4ccccc4)c5ccccc5)c6ccccc6)c7ccccc7
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C41H29NO/c43-41-37(31-18-8-2-9-19-31)28-36(29-38(41)32-20-10-3-11-21-32)42-39(33-22-12-4-13-23-33)26-35(30-16-6-1-7-17-30)27-40(42)34-24-14-5-15-25-34/h1-29H
Formula Weight 551.2249
Chemical Name or Material Reichardt's Dye (Technical Grade)