missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65, p Ser276 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
320.00€ - 482.00€
Specifications
| Antigen | RelA/NFkB p65 (p Ser276) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200 Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260314
|
Novus Biologicals
NB100-82086-0.025ML |
0.025 mL |
320.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18741104
|
Novus Biologicals
NB100-82086 |
0.05 mL |
482.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RelA/NFkB p65 (p Ser276) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200 Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| Q04206 | |
| 5970 | |
| IgG | |
| Affinity Purified | |
| 65 kDa |
| 1.0 mg/mL | |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human | |
| MGC131774, nf kb p65, NF kB p65 phospho, NFkB p65, NF-kB p65, NFkB p65 phospho, NF-kB p65 phospho, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, phospho NF kB p65, phospho NFkB p65, phospho NF-kB p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| RELA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title