missing translation for 'onlineSavingsMsg'
Learn More

Relpax, TRC

Product Code. 30416710
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416710 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416710 Supplier Toronto Research Chemicals Supplier No. TRCR1434005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Relpax
CAS 177834-92-3
Molecular Formula C22 H26 N2 O2 S . Br H
Quantity 5 mg
Synonym 3-[[(2R)-1-methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-Indole hydrobromide (1:1), 1H-Indole','3-[(1-methyl-2-pyrrolidinyl)methyl]-5-[2-(phenylsulfonyl)ethyl]-','monohydrobromide','(R)-, 1H-Indole','3-[[(2R)-1-methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-','monohydrobromide (9CI), (R)-5-[2-(Benzenesulfonyl)ethyl]-3-[(N-methylpyrrolidin-2-yl)methyl]-1H-indole hydrobromide, Eletriptan hydrobromide, Relert, Relpax, UK 116044-04
InChI Formula InChI=1S/C22H26N2O2S.BrH/c1-24-12-5-6-19(24)15-18-16-23-22-10-9-17(14-21(18)22)11-13-27(25,26)20-7-3-2-4-8-20;/h2-4,7-10,14,16,19,23H,5-6,11-13,15H2,1H3;1H/t19-;/m1./s1
SMILES Br.CN1CCC[C@@H]1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23
IUPAC Name 5-[2-(benzenesulfonyl)ethyl]-3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-1H-indole;hydrobromide
Molecular Weight (g/mol) 463.43
Formula Weight 462.0977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.