Learn More
REN Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29330-1-AP-20UL
 View more versions of this product
Product Code. 18078085
  • 145.00€ / 20µL
Description
The REN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human REN. This antibody recognizes human, monkey antigen. The REN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
REN
500 μg/mL
Rabbit
Primary
Human, Monkey
150 kDa
IgG
Western Blot, ELISA
Angiotensinogenase, FLJ10761, REN, renin
20 μL
5972
Store at -20°C. Stable for one year after shipment.