Learn More
Rephile™ RephiSolo U Pack IX kit
Product Code. 15439749 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15439749 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15439749
  • ONLINE EXCLUSIVE PRICE
    474.99€
    386.11€ / Pack of 1
    Save 88.88€
    18% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is designed for removing ionic and organic contaminants below trace level to match the water quality required for your application.
RephiSolo U Pack IX kit for MilliQ Century™ systems. Comparable to QTUMMPEIX.
Compatible with following Millipore™ systems:
  • Milli-Q Academic™
  • Milli-Q Biocel™
  • Milli-Q Element™
  • Milli-Q Gradient™
  • Milli-Q Synthesis™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit