Learn More
Eppendorf™ Replacement Rotor Lids
Product Code. 15158354 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
F-35-6-30 Rotor
FA-45-12-17 Rotor
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15158354 FA-45-12-17 Rotor Each
10435325 F-35-6-30 Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15158354 Supplier Eppendorf™ Supplier No. 5409701002
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Designed for use with Eppendorf Rotors
  • Replacement Rotor Lids
  • Available for a wide range of products
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
For Use With (Equipment) FA-45-12-17 Rotor
Color Black
Description Rotor lid for FA-45-12-17, aerosol-tight
Product Type Aerosol-tight