Learn More
Veolia Water Systems Resin Cartridge
Product Code. 11964887
Change view
Click to view available options
Quantity:
2 Pack
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11964887 2 Pack Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11964887 Supplier Veolia Water Systems Supplier No. MC:DS
  • ONLINE EXCLUSIVE PRICE
    667.80€
    522.08€ / Pack of 2
    Save 145.72€
    21% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Resin cartridge used to remove ionic contaminants to achieve specified water quality.
  • Removes ionic contaminants to achieve specified water quality
  • ELGA cartridges and accessories
  • X2 resin cartridge for Micromeg
Specifications
For Use With (Equipment) For Micromeg
Quantity 2 Pack
Type Cartridge