missing translation for 'onlineSavingsMsg'
Learn More

Respiratory Syncytial Virus Glycoprotein F Antibody (342), Novus Biologicals™

Rabbit Monoclonal Antibody

201.00€ - 452.00€

Specifications

Antigen Respiratory Syncytial Virus Glycoprotein F
Clone 342
Dilution ELISA 1:5000-1:10000, Lateral Flow Assay 1 mg/mL
Applications ELISA, Lateral Flow
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30281309
View Documents
Novus Biologicals
NBP3-43763-20ul
20 μL
201.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30281308
View Documents
Novus Biologicals
NBP3-43763-100ul
100 μL
452.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for ELISA,Lateral Flow
Specifications

Specifications

Respiratory Syncytial Virus Glycoprotein F
ELISA 1:5000-1:10000, Lateral Flow Assay 1 mg/mL
Monoclonal
Purified
RUO
0.2 μm filtered solution in PBS
This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (Uniprot#: P03420-1; Met 1-Thr 529).
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
342
ELISA, Lateral Flow
Unconjugated
Rabbit
Virus
Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.