missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret Antibody (132507), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB718F
This item is not returnable.
View return policy
Description
Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Ret | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 132507 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Ret/Fc Chimera, Leu29-Arg635, Accession # P07949 | |
| 0.1 mL | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 5979 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction