Learn More
Applied Biosystems™ 3500 Dx Series 96-Well Fast (0.1 mL) Retainer & Base Set
Product Code. 17296483 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Sets
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17296483 4 Sets Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17296483 Supplier Applied Biosystems™ Supplier No. 4410229
  • ONLINE EXCLUSIVE PRICE
    365.00€
    270.10€ / Pack of 4
    Save 94.90€
    26% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
This is a retainer and base set (fast) designed for septa retention in 8-tube strip and 96-well plate formats for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only.
  • This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
TRUSTED_SUSTAINABILITY
Specifications
Compatibility 96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
Content And Storage Ambient storage.
Each package contains 4 sets.
Format 96-well
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Retainer and Base Set
Quantity 4 Sets
Shipping Condition Room Temperature
For In Vitro Diagnostic Use Only