missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144688
This item is not returnable.
View return policy
Description
Immunogen sequence: GKPVAVTSNR GTGRAPLRPG ANPQLEWQYY QNKILKGKAA DIPDSSLWEN PDSAQANGID SVLSRAEIAS CSYEARQLGI KNGMFFGHAK QLCPNLQAVP YDFHAYKEVA QTLYETLASY THNIEAVSCD EA Antigen sequence identity: Mouse 88%; Rat 92%.
This gene encodes a protein with similarity to the S. cerevisiae mutagenesis protein Rev1. The Rev1 proteins contain a BRCT domain, which is important in protein-protein interactions. A suggested role for the human Rev1-like protein is as a scaffold that recruits DNA polymerases involved in translesion synthesis (TLS) of damaged DNA. Two alternatively spliced transcript variants that encode different proteins have been found.
Specifications
| REV1 | |
| Polyclonal | |
| Unconjugated | |
| Rev1 | |
| 1110027I23Rik; AIBP80; alpha integrin-binding protein 80; AU022044; DNA repair protein REV1; REV1; REV1 homolog; REV1 homolog (S. cerevisiae); REV1- like; REV1, DNA directed polymerase; REV1, polymerase (DNA directed); REV1L; rev1-like terminal deoxycytidyl transferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBZ9 | |
| Rev1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 441-572. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction