missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00€ - 662.00€
Specifications
| Antigen | RFP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203511
|
Novus Biologicals
NBP1-45840-0.025MG |
0.025 mg |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18225761
|
Novus Biologicals
NBP1-45840-0.05MG |
0.05 mg |
409.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18287172
|
Novus Biologicals
NBP1-45840 |
0.1 mg |
662.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RFP Polyclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot.Specifications
| RFP | |
| Polyclonal | |
| Rabbit | |
| dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Non-species specific | |
| Recombinant RFP expressed from E. coli. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title