missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals
This item is not returnable.
View return policy
Description
RFP Polyclonal antibody specifically detects RFP in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Specifications
| RFP | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:200-1:2000, ELISA 1:28700-1:48700, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready | |
| DSRED, GFP-like chromoprotein, Red Fluorescent Protein, RFP | |
| The immunogen is a Red Fluorescent Protein (RFP) fusion protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma. | |
| 100 μg | |
| Zinc Finger | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction