Learn More
Invitrogen™ RGMA Recombinant Rabbit Monoclonal Antibody (24GB1320)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557221
 View more versions of this product
Product Code. 30276196
  • 500.00€ / 100µL
Description
RGMA Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Member of the repulsive guidance molecule (RGM) family that performs several functions in the developing and adult nervous system. Regulates cephalic neural tube closure, inhibits neurite outgrowth and cortical neuron branching, and the formation of mature synapses. Binding to its receptor NEO1/neogenin induces activation of RHOA-ROCK1/Rho-kinase signaling pathway through UNC5B-ARHGEF12/LARG-PTK2/FAK1 cascade, leading to collapse of the neuronal growth cone and neurite outgrowth inhibition. Furthermore, RGMA binding to NEO1/neogenin leads to HRAS inactivation by influencing HRAS-PTK2/FAK1-AKT1 pathway. It also functions as a bone morphogenetic protein (BMP) coreceptor that may signal through SMAD1, SMAD5, and SMAD8.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGMA
Recombinant Monoclonal
0.67 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PCX7, Q96B86
RGMA
A synthesized peptide derived from human RGMA (aa 401-435).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1320
Unconjugated
RGMA
BC059072; C230063O06; repulsive guidance molecule; repulsive guidance molecule A; repulsive guidance molecule family member a; RGM; RGM domain family member A; RGM domain family, member A; RGMA
Rabbit
Affinity chromatography
RUO
244058, 308739, 56963
-20°C