missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGMA Recombinant Rabbit Monoclonal Antibody (24GB1320)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557222
This item is not returnable.
View return policy
Description
RGMA Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Member of the repulsive guidance molecule (RGM) family that performs several functions in the developing and adult nervous system. Regulates cephalic neural tube closure, inhibits neurite outgrowth and cortical neuron branching, and the formation of mature synapses. Binding to its receptor NEO1/neogenin induces activation of RHOA-ROCK1/Rho-kinase signaling pathway through UNC5B-ARHGEF12/LARG-PTK2/FAK1 cascade, leading to collapse of the neuronal growth cone and neurite outgrowth inhibition. Furthermore, RGMA binding to NEO1/neogenin leads to HRAS inactivation by influencing HRAS-PTK2/FAK1-AKT1 pathway. It also functions as a bone morphogenetic protein (BMP) coreceptor that may signal through SMAD1, SMAD5, and SMAD8.
Specifications
| RGMA | |
| Recombinant Monoclonal | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PCX7, Q96B86 | |
| RGMA | |
| A synthesized peptide derived from human RGMA (aa 401-435). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1320 | |
| Unconjugated | |
| RGMA | |
| BC059072; C230063O06; repulsive guidance molecule; repulsive guidance molecule A; repulsive guidance molecule family member a; RGM; RGM domain family member A; RGM domain family, member A; RGMA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 244058, 308739, 56963 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction