missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97095UV
This item is not returnable.
View return policy
Description
RGS1 Polyclonal specifically detects RGS1 in Human samples. It is validated for ELISA.
Specifications
| RGS1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 350 | |
| 1R20Early response protein 1R20, BL34B-cell activation protein BL34, IER1, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1 | |
| Produced in rabbits immunized with purified, recombinant RGS1 (NP_002913.3; Met1-Lys196). | |
| 0.1 mL | |
| Neuroscience, Neurotransmission, Signal Transduction | |
| 5996 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction