Learn More
Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors
Product Code. 10384423 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Continuous Flow
Continuous Flow, Position Sensing Switch
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10384423 Continuous Flow, Position Sensing Switch Each
10171033 Continuous Flow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10384423 Supplier Thermo Scientific™ Supplier No. 7725I
Estimated Shipment: 11-01-2027
Add to basket
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Ensure accurate, precise HPLC injections and protect against pressure shock with the dual-mode Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors.
Maintain continuous HPCL flow between the load and inject positions and protect against pressure shock by using Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors. The Make-Before-Break (MBB) design ensures uninterrupted flow during sample injection. These stainless-steel, dual-mode injectors can use partial filling for zero sample waste or complete filling for better reproducibility. Samples from 1μL to 5mL can be injected with high accuracy and precision. The 7725i model features a position-sensing switch for a reproducible start signal.
  • Stainless steel construction
  • Make-Before-Break (MBB) design
  • Can use partial filling for zero sample waste or complete filling for better reproducibility
  • Inject 1μL to 5mL with high accuracy and precision
  • 7725i features a position sensing switch for a reproducible start signal
(Includes: 20μL sample loop)
TRUSTED_SUSTAINABILITY
Spécification
Mode Dual Mode
Volume (Metric) Sample 20 μL
Description Continuous Flow, Position Sensing Switch
For Use With (Equipment) HPLC
Type Continuous Flow HPLC Sample Injector
Stator Material Stainless Steel
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.