Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Product Code. 10161263
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector 7030
Rheodyne Injector 8125
Rheodyne Injector 9010
Rheodyne Injector 9125
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7000/7010/7040
Rheodyne Injectors 7000/7010/7040/7067
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7060/7066/9060
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
PEEK
Vespel
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material unitSize
10161263 Rheodyne Injectors 7000/7010/7040/7067 Vespel Each
10290285 Rheodyne Injectors 3725/3725i/3725-038/3725i-038 PEEK Each
10445163 Rheodyne Injectors 7000/7010/7040 ETFE Each
10671296 Rheodyne Injector 7030 Vespel Each
10576005 Rheodyne Injectors 7060/7066 Vespel Each
10648705 Rheodyne Injectors 7060/7066/9060 ETFE Each
10150893 Rheodyne Injectors 7125/7126 Vespel Each
10269514 Rheodyne Injectors 7125/7126 ETFE Each
10427534 Rheodyne Injectors 8125/8126 Vespel Each
10510126 Rheodyne Injector 8125 ETFE Each
10179464 Rheodyne Injector 9010 ETFE Each
10280845 Rheodyne Injector 9125 ETFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10161263 Supplier Thermo Scientific™ Supplier No. 7010039
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Rotor Seals
Material Vespel
Description 7010-039
For Use With (Equipment) Rheodyne Injectors 7000/7010/7040/7067
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.