Learn More
RHOH Monoclonal Antibody (OTI9F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809996S
Description
RHOH Monoclonal antibody specifically detects RHOH in Human samples. It is validated for Western BlotSpecifications
| RHOH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P83369 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 134353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9F4 | |
| Unconjugated | |
| LSM11 | |
| 5830400A04Rik; ARHH; AU019774; GTP-binding protein TTF; ras homolog family member H; ras homolog gene family, member H; RHOH; rho-related GTP-binding protein RhoH; Translocation three four protein; TTF; TTF, translocation three four | |
| Full length human recombinant protein of human LSM11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.