missing translation for 'onlineSavingsMsg'
Learn More

Ribonucleic acid, MP Biomedicals™

Product Code. 11434321 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434321 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434321 Supplier MP Biomedicals Supplier No. 0215202680

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 63231-63-0
MDL Number MFCD00132196
Synonym RNA
IUPAC Name Ribonucleic acid

Specifications

Color White
Quantity 100 g
Physical Form Powder
Chemical Name or Material Ribonucleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.