Learn More
Ribosomal protein L4 Mouse anti-Human, Unconjugated, Clone: 3C7A8, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Ribosomal protein L4 |
|---|---|
| Clone | 3C7A8 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889405
|
Proteintech
67028-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879405
|
Proteintech
67028-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Ribosomal protein L4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Ribosomal protein L4. This antibody recognizes human, mouse, rat antigen. The Ribosomal protein L4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Ribosomal protein L4 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 6124 | |
| Primary |
| 3C7A8 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.