missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosome maturation protein SBDS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85646-25ul
This item is not returnable.
View return policy
Description
Ribosome maturation protein SBDS Polyclonal antibody specifically detects Ribosome maturation protein SBDS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Ribosome maturation protein SBDS | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PTNQIRLTNVAVVRMKRAGKRFEIACYKNKVVGWRSGVEKDLDEVLQTHSVFVNVSKGQVAKKEDLISAFGTDDQTEICKQILTKGEVQVSDKERHTQLEQMFRDIATIVADKCVNPETKRPYTVILIERAMKDIHYSVK | |
| 25 μL | |
| metabolism | |
| 51119 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction